Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific™ FitBag™ UVM1 Broth

Shorten your workflow with easy to prepare 2.7L, 4.5L or 9L media bags: ready in 5 to 10 minutes, while segmented structure allows pre-filling.

$929.00 - $981.00

Specifications

Description FitBag UVM1 Broth
Storage Requirements 10-30°C
Type Media
Format Convenience Format
View More Specs

Products 3
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
DF0863A View Documents Thermo Scientific™
DF0863A
30 x 2.7L Each for $945.50
Add to cart
 
DF0863C View Documents Thermo Scientific™
DF0863C
10 x 9.0L Each for $929.00
Add to cart
 
DF0863B View Documents Thermo Scientific™
DF0863B
20 x 4.5L Each for $981.00
Add to cart
 
Description

Description

Save time in preparing a selective enrichment medium that conforms to the USDA-FSIS method for the isolation of Listeria monocytogenes, using Thermo Scientific™ FitBag™ UVM1 Broth. The irradiated medium is supplied in lightweight, transparent, plastic bags—just add water and the medium is ready for use. The bag’s segmented structure allows pre-filling so that you can reconstitute in two minutes when needed. Available in 2.7L, 4.5L or 9L sizes to suit your workflow, medium can be dispensed through the outlet tube as required.

Shorten your workflow with FitBag UVM1 Broth. FitBag Enrichment Media is quick, easy-to-prepare, and highly adaptable to laboratory workflows and automated equipment, such as dilutors. No further supplementation is required.

Cost-effective: no need for weighing of media or sterilization, reducing the burden of media preparation, which frees up resources and equipment.

Flexible: segmented structure allows advanced preparation by filling one segment with deionized water from which FitBag Enrichment Media can be reconstituted within two minutes. Pre-sterilized water may also be added under aseptic conditions.

Easy-to-use: fill and dispense with minimum training.

Energy saving:
gamma irradiated, eliminating the need for costly thermal processing.

Safe: eliminate handling of glassware, weighing of culture media, and the need to add supplementary reagents. Empty bags can be disposed of with normal laboratory waste.

Reduces waste: unprepared FitBag Enrichment Media can be kept for up to 24 months. Rehydrated media has a shelf life of 10 days.

Quality guaranteed: compliant with ISO 11133 and ISO 7218. Quality certificates are available for every batch, reducing the QC testing needed.

Space-saving:
conveniently packed in flat boxes, FitBag Enrichment Media takes up relatively little space compared to prepared media and can be made up as required.

Specifications

Specifications

FitBag UVM1 Broth
10-30°C
Media
Convenience Format
Safety and Handling

Safety and Handling

ShelfLife : 2 years (rehydrated 10 days)

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit