Learn More
BiolegendSupplier Diversity Partner FITC anti-human HLA-DR, DP, DQ  DFS Item
Manufacturer:  BiolegendSupplier Diversity Partner 361706
FITC anti-human HLA-DR, DP, DQ [Tü39]; Isotype: Mouse IgG2a, κ; Reactivity: Human, Cross-Reactivity: Baboon, Cynomolgus, Cat (Feline), Dog (Canine), Cattle; Apps: FC; Size: 100 tests
Catalog No. 50-712-204