Learn More
Cole-Parmer Masterflex Fitting, Nylon, Straight, Hose Barb to Threaded Adapter, 3/4" ID x 1/2" NPT(M); 10/PK  DFS Item
Manufacturer:  Cole-Parmer 3062172
Available in a variety of materials and configurations to meet your need. Adapts from a NPT male thread to a hose barb.
Catalog No. 50-629-025