Learn More
Cole-Parmer Masterflex Fitting, Nylon, Straight, Hose Barb to Threaded Adapter, 5/8" ID x 3/4" NPT(M); 10/PK  DFS Item
Manufacturer:  Cole-Parmer 3062110
Available in a variety of materials and configurations to meet your need. Adapts from a NPT male thread to a hose barb.
Catalog No. 50-628-719