Learn More
Cole-Parmer Masterflex Fitting, Nylon, Elbow, Hose Barb to Threaded Adapter, 3/8" ID x 3/8" NPT(M); 10 PK  DFS Item
Manufacturer:  Cole-Parmer 3062305
Barbed elbow fittings are designed for quick and easy assembly without tools. Combine these simple and economical fittings with our flexible tubing for use in your low-pressure applications (generally less than 30 psi).
Catalog No. 50-629-985