Learn More
Cole-Parmer Masterflex Fitting Kit, PVDF, Straight/Reducer/Plug, Hose Barb, Small Fittings; 1 Set  DFS Item
Manufacturer:  Cole-Parmer 3151404
Organize your workstation with these convenient fitting kits. Assorted fittings come in impact resistant plastic box with secure latch to keep fittings organized and in place. Label compartments make fitting location quick and easy. Choose from four materials: high-density polyethylene (HDPE), black nylon, natural polypropylene, and natural PVDF.
Catalog No. 50-629-627