Learn More
Cole-Parmer CPC (Colder) LC22004 Inserts; hose barb straight-through fitting; 1/4" ID; 1.35" length  DFS Item
Manufacturer:  Cole-Parmer 3140345
Metal quick-disconnect couplings require a fitting body and an insert with the same flow size (order separately). Withstand vacuum to 250 psi, temperatures from -40 to 180°F, and can be used with gases or liquids.
Catalog No. 50-627-752