Learn More
Cole-Parmer Masterflex Fitting, Nylon, Straight, Hose Barb Reducer, 1" ID x 3/4" ID; 50/PK  DFS Item
Manufacturer:  Cole-Parmer 3062614
These economical straight barbed reducing fittings easily connect fluid paths without the use of tools. Just slip your flexible tubing or hose onto the barb to create a secure seal.
Catalog No. 50-629-386