Learn More
Cole-Parmer Masterflex Fitting, HDPE, Straight, Hose Barb Union, 5/16" ID; 10/PK  DFS Item
Manufacturer:  Cole-Parmer 3062187
These economical straight barbed fittings easily connect fluid paths without the use of tools. Just slip your flexible tubing or hose onto the barb to create a secure seal.
Catalog No. 50-629-233