Learn More
Cole-Parmer CPC (Colder) MPC17006T39 Quick-Disconnect Body, PS, 3/8" ID; 5/Pk  DFS Item
Manufacturer:  Cole-Parmer 3131122
All-plastic quick-disconnect couplings require a standard fitting body and standard insert or large fitting body and large insert (order separately). Simply press the thumb latch to disconnect fitting halves. Available in polycarbonate (PC), Acetal, and polysulfone (PSF).
Catalog No. 50-627-872