Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
FIV gp95 Antibody (SU1-30), HRP, Novus Biologicals™
Mouse Monoclonal Antibody
Supplier: Novus Biologicals NB10064378H
Description
FIV gp95 Monoclonal antibody specifically detects FIV gp95 in Virus samples. It is validated for Western BlotSpecifications
FIV gp95 | |
Monoclonal | |
HRP | |
Mouse | |
Protein A purified | |
RUO | |
Virus | |
Purified |
Western Blot | |
SU1-30 | |
PBS | |
Made to FIV gp95 | |
0.125 mL | |
Primary | |
Store at 4°C in the dark. | |
IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Product Content Correction