Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Diversified Spaces™ Fixed Height Riser Table
Riser with 10 in. clearance above work surface.
Supplier: Diversified Spaces™ PR7132K30N
Description
- Riser with chemical-resistant finish
Specifications
Fixed Height Riser Table | |
30 x 54 in. | |
ChemGuard |
12 in. D riser | |
Oak Apron |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction