Learn More
Fischer Technical Company™ Fixed Volume Mini-Pipettors Set
These fixed volume mini pipettors conveniently come packed with 5 pipettors and a pack of 1000 tips.
$189.00
Specifications
Autoclavable | Autoclavable |
---|---|
Material | Plastic |
Sterility | Non-sterile |
Graduated | No |
Length (Metric) | 9 cm |
Description
This kit comes with five fixed volume mini-pipettors and a pack of 1000 pipette tips.
- Unique miniature pipettors provide accurate fixed volume dispensing
- Ideal for micro-volumes and gel loading applications
- Accepts universal fit, disposable pipette tips
Specifications
Autoclavable | |
Non-sterile | |
9 cm | |
No |
Plastic | |
No | |
Mini Pipettor | |
Liquid Handling |