Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

FIZZ1/RELM alpha Rat anti-Mouse, Alexa Fluor™ 350, Clone: RM0313-9A39, Novus Biologicals™

Rat Monoclonal Antibody

Manufacturer:  Novus Biologicals NBP212440AF350

Catalog No. NP212440A30


Add to cart

Description

Description

FIZZ1/RELM alpha Monoclonal antibody specifically detects FIZZ1/RELM alpha in Mouse samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin).
Specifications

Specifications

FIZZ1/RELM alpha
RM0313-9A39
Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin
Conjugated, Purified
RETNLA
Mouse
Protein G purified
Store at 4°C in the dark.
Monoclonal
Mouse
Immunohistochemistry, Immunohistochemistry (Paraffin), Western Blot
Alexa Fluor™ 350
50 mM sodium borate with 0.05% sodium azide
1810019L16Rik, Fizz1, Fizz-1, HIMF, RELMa, RELMalpha, RELM-alpha, Retnla resistin like alpha, Xcp2
Rat
IgG2
0.1 mL
Primary
57262.0
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit