Learn More
FIZZ1/RELM alpha Rat anti-Mouse, Alexa Fluor™ 594, Clone: RM0313-9A39, Novus Biologicals™
Rat Monoclonal Antibody
Manufacturer:  Novus BiologicalsSupplier Diversity Partner NBP212440AF594
Catalog No. NP212440A94
Add to cart
Description
Description
FIZZ1/RELM alpha Monoclonal antibody specifically detects FIZZ1/RELM alpha in Mouse samples. It is validated for Western Blot,Immunohistochemistry,Immunohistochemistry (Paraffin).
Specifications
Specifications
FIZZ1/RELM alpha
RM0313-9A39
Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin
1810019L16Rik, Fizz1, Fizz-1, HIMF, RELMa, RELMalpha, RELM-alpha, Retnla resistin like alpha, Xcp2
Rat
IgG2
Store at 4°C in the dark.
Monoclonal
Mouse
Immunohistochemistry, Immunohistochemistry (Paraffin), Western Blot
Alexa Fluor™ 594
50 mM sodium borate with 0.05% sodium azide
RETNLA
Mouse
0.1 mL
Primary
57262