Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ FJX1 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA542965
Description
Peptide sequence: MVALERGGCG RSSNRLARFA DGTRACVRYG INPEQIQGEA LSYYLARLLG Sequence homology: Dog: 79%; Guinea Pig: 100%; Human: 100%; Mouse: 100%; Rat: 100%.
FJX1 is the human ortholog of mouse and Drosophila four-jointed gene product. The Drosophila protein is important for growth and differentiation of legs and wings, and for proper development of the eyes. The exact function of FJX1 gene in humans is not known..The protein encoded by this gene is the human ortholog of mouse and Drosophila four-jointed gene product. The Drosophila protein is important for growth and differentiation of legs and wings, and for proper development of the eyes. The exact function of this gene in humans is not known.
Specifications
FJX1 | |
Polyclonal | |
Unconjugated | |
FJX1 | |
FJX1; FLJ22416; FLJ25593; four jointed box 1; four jointed box 1 (Drosophila); four-jointed box kinase 1; four-jointed box protein 1; Four-jointed protein homolog; four-jointed protein x1; LOW QUALITY PROTEIN: four-jointed box protein 1; putative secreted ligand homologous to fjx1 | |
Rabbit | |
Affinity chromatography | |
RUO | |
24147 | |
-20°C, Avoid Freeze/Thaw Cycles | |
Liquid |
Western Blot | |
0.5 mg/mL | |
PBS with 2% sucrose and 0.09% sodium azide | |
Q86VR8 | |
FJX1 | |
Synthetic peptide directed towards the N-terminal of human FJX1 (aa 153-202). | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction