Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
FJX1 Rabbit anti-Human, Rat, Polyclonal, Novus Biologicals™
Rabbit Polyclonal Antibody
$204.00 - $482.50
Specifications
Antigen | FJX1 |
---|---|
Applications | Western Blot |
Classification | Polyclonal |
Conjugate | Unconjugated |
Form | Purified |
Description
FJX1 Polyclonal specifically detects FJX1 in Human, Rat samples. It is validated for Western Blot.Specifications
FJX1 | |
Polyclonal | |
Purified | |
RUO | |
PBS with 50% glycerol, pH7.3. | |
24147 | |
IgG | |
Affinity purified |
Western Blot | |
Unconjugated | |
Rabbit | |
Human, Rat | |
FLJ22416, FLJ25593, four jointed box 1 (Drosophila), four-jointed box protein 1, Four-jointed protein homolog, putative secreted ligand homologous to fjx1 | |
Recombinant fusion protein containing a sequence corresponding to amino acids 138-437 of human FJX1 (NP_055159.2). | |
Primary | |
Store at -20°C. Avoid freeze-thaw cycles. |
Spot an opportunity for improvement?
Provide Content Correction
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title