Learn More
Selleck Chemical LLCSupplier Diversity Partner Tacrolimus (FK506) 100mg 104987-11-3 FR900506, Fujimycin, Prograf  DFS Item
Manufacturer:  Selleck Chemical LLCSupplier Diversity Partner S5003100MG
Tacrolimus (FK506, FR900506, Fujimycin, Prograf) is a 23-membered macrolide lactone, it reduces peptidyl-prolyl isomerase activity in T cells by binding to the immunophilin FKBP12 (FK506 binding protein) creating a new complex. Tacrolimus also inhibits the phosphatase activity of calcineurin. Tacrolimus induces vascular endothelial autophagy.
Catalog No. 50-648-4