Learn More
FK 866 hydrochloride (Visfatin), Tocris™
Non-competitive and potent inhibitor of NAMPT (nicotinamide phosphoribosyltransferase, visfatin, PBEF1) (Ki = 0.3nM); inhibits NAD biosynthesis. Induces delayed cell death by apoptosis in HepG2 human
Manufacturer:  R&D Systems 4808/50
Quantity 50mg
 View more versions of this product
Catalog No. 48-085-0
  Check Availability
Add to cart
Specifications Description & Specifications
Chemical Identifiers
C24H29N3O2·HCl
427.97
2-(E)-N-[4-(1-Benzoyl-4-piperidinyl)butyl]-3-(3-pyridinyl)-2-propenamide hydrochloride
FK 866 hydrochloride (Visfatin)
Specifications
50mg
427.97
FK 866 hydrochloride (Visfatin)
C24H29N3O2·HCl
2-(E)-N-[4-(1-Benzoyl-4-piperidinyl)butyl]-3-(3-pyridinyl)-2-propenamide hydrochloride
Safety and Handling
Recommended Storage : Store at -20°C