Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

FKBP12 Mouse anti-Human, PE-Cyanine7, Clone: OTI3B3, Novus Biologicals™

Mouse Monoclonal Antibody

Manufacturer:  Novus Biologicals NBP270741PECY7

Catalog No. N270741PEC7


Add to cart

Description

Description

FKBP12 Monoclonal antibody specifically detects FKBP12 in Human samples. It is validated for Flow Cytometry.
Specifications

Specifications

FKBP12
OTI3B3
Flow Cytometry
Affinity Purified, Conjugated
FKBP1A
Full length recombinant protein of human FKBP12 (NP_000792) produced in HEK293T cells.
Immunogen affinity purified
Autophagy, Cancer, mTOR Pathway
Primary
2280.0
Flow Cytometry
PE-Cyanine7
PBS with 0.05% Sodium Azide
12 kDa FKBP, EC 5.2.1.8, FK506 binding protein 1A, 12kDa, FK506-binding protein 1A, FKBP1, FKBP12C, FKBP12FKBP12-Exip3, FKBP-12T-cell, 12-kD, FKBP-1A, peptidyl-prolyl cis-trans isomerase FKBP1A, PPIase FKBP1A, protein kinase C inhibitor 2, rotamase
Mouse
IgG2b
0.1 mL
Store at 4°C in the dark. Do not freeze.
Monoclonal
Human
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit