Learn More
Abnova™ FKBP14 293T Cell Transient Overexpression Lysate (Denatured)
Human overexpression lysate
Manufacturer:  Abnova™ H00055033T01
Catalog No. 89-944-601
Add to cart
Specifications
Specifications
Western Blot
293T
NM_017946.2
pCMV-FKBP14 full-length
100 uL
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
Human
FKBP14 293T Cell Transient Overexpression Lysate(Denatured)
Theoretical M.W. 23.32kDa
Transient overexpression cell lysate was tested with anti-FKBP14 antibody (Mfr. No. H00055033-B01) by Western Blot.
1X Sample Buffer (50 mM Tris-HCl, 2% SDS, 10% glycerol, 300 mM 2-mercaptoethanol, 0.01% Bromophenol blue)