Learn More
Aviva Systems Biology Corp FKBP2 Antibody - N-terminal region (ARP58465_P050) | 100uL
Supplier:  Aviva Systems Biology Corp ARP58465_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against FKBP2. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the N terminal region of human FKBP2. Alternate names: FKBP-13, PPIaseApplicationWB. Species reactivity: Cow, Dog, Goat, Guinea Pig, Horse, Human, Mouse, Rat, Zebrafish
Catalog No. 50-014-62315