Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ FKBP4 Monoclonal Antibody (Hi52C), FITC
Mouse Monoclonal Antibody
Supplier: Invitrogen™ MA545164
This item is not returnable.
View return policy
Description
0.5 μg/mL was sufficient for detection of FKBP52 in 20 μg total protein using WB by colorimetric immunoblot analysis using Goat Anti-Mouse IgG:HRP as the secondary. Detects approximately 52kDa. Heavy chain migrates close to FKBP52 on SDS PAGE.
The function of this protein remains unknown.
Specifications
FKBP4 | |
Monoclonal | |
1 mg/mL | |
PBS, 9.1mM sodium bicarbonate with 640.91mM DMSO, 136.36mM ethanolamine and no preservative | |
P30416, Q02790, Q9QVC8 | |
FKBP4 | |
Synthetic peptide corresponding to the residues of human FKBP52. | |
100 μg | |
Primary | |
Human, Mouse, Rat, Canine, Hamster | |
Antibody | |
IgG2a |
Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot, Immunocytochemistry | |
Hi52C | |
FITC | |
FKBP4 | |
51 kDa FK506-binding protein; 52 kDa FK506-binding protein; 52 kDa FKBP; 59 kDa immunophilin; 59kDa; AL022792; AW208983; FK506 binding protein 4; FK506 binding protein 4, 59kDa; FK506 binding protein 52; FK506-binding protein 4; FK506-binding protein 4 (59kD); FKBP prolyl isomerase 4; Fkbp4; FKBP-4; FKBP51; FKBP52; FKBP-52; FKBP52 protein; FKBP59; Fkpb52; HBI; HSP binding immunophilin; Hsp56; hsp90 binding protein; HSP-binding immunophilin; immunophilin FKBP52; p52; P59; p59 protein; peptidylprolyl cis-trans isomerase; peptidyl-prolyl cis-trans isomerase FKBP4; Peptidyl-prolyl cis-trans isomerase FKBP4, N-terminally processed; PPIase; PPIase FKBP4; rotamase; T-cell FK506-binding protein, 59kD | |
Mouse | |
Protein G | |
RUO | |
100762395, 14228, 2288, 260321, 477726 | |
4°C, store in dark | |
Liquid |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction