Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

FKBP51/FKBP5 Mouse anti-Human, Alexa Fluor™ 405, Clone: OTI3E9, Novus Biologicals™

Mouse Monoclonal Antibody

Manufacturer:  Novus Biologicals NBP270742AF405

Catalog No. N270742A405


Add to cart

Description

Description

FKBP51/FKBP5 Monoclonal antibody specifically detects FKBP51/FKBP5 in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence.
Specifications

Specifications

FKBP51/FKBP5
OTI3E9
Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence
Affinity Purified, Conjugated
FKBP5
Full-length protein expressed in 293T cell transfected with human FKBP51/FKBP5 expression vector.
Immunogen affinity purified
Cancer
Primary
2289.0
Immunocytochemistry, Immunofluorescence, Immunohistochemistry, Western Blot
Alexa Fluor 405
50mM Sodium Borate with 0.05% Sodium Azide
51 kDa FKBP, 54 kDa progesterone receptor-associated immunophilin, AIG6,51 kDa FK506-binding protein, Androgen-regulated protein 6, FF1 antigen, FK506 binding protein 5, FK506-binding protein 5, FKBP-5, FKBP-51, FKBP51PPIase FKBP5, FKBP54EC 5.2.1.8, HSP90-binding immunophilin, MGC111006, p54, P54,51 kDa FK506-binding protein 5, peptidylprolyl cis-trans isomerase, peptidyl-prolyl cis-trans isomerase FKBP5, PPIase, Ptg-10, rotamase, T-cell FK506-binding protein
Mouse
IgG1
0.1 mL
Store at 4°C in the dark.
Monoclonal
Human
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit