Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

FKBP7 Rabbit anti-Human, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Manufacturer:  Invitrogen PA548472

Catalog No. PA548472

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.


Description

Description

The protein encoded by this gene belongs to the FKBP-type peptidyl-prolyl cis/trans isomerase family. Members of this family exhibit PPIase activity and function as molecular chaperones. A similar protein in mouse is located in the endoplasmic reticulum and binds calcium.
Specifications

Specifications

FKBP7
Polyclonal
PBS with 0.09% sodium azide
Q9Y680
Peptidyl-prolyl cis-trans isomerase FKBP7, PPIase FKBP7, 23 kDa FK506-binding protein, 23 kDa FKBP, FKBP-23, FK506-binding protein 7, FKBP-7, Rotamase, FKBP7, FKBP23
Rabbit
IgG
400 μL
-20° C, Avoid Freeze/Thaw Cycles
Primary
51661
Flow Cytometry, Immunohistochemistry (Paraffin), Western Blot
Unconjugated
FKBP7
Liquid
FKBP7
KLH conjugated synthetic peptide between 195-224 amino acids from the C-terminal region of human FKBP7.
Antigen affinity chromatography
RUO
Antibody
Polyclonal
Human
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit