Learn More
White Knight™ Flame-Resistant Unisex Lab Coat
Designed to provide a superior level of protection against flash fire, electric arc (HRC 1), and other dangerous conditions. This Flame-Resistant lab coat has numerous applications in both laboratory and production settings.
$175.00 - $275.00
Specifications
Arc Flash Rating HRC 1
Certifications/Compliance NFPA 2112 Certified Garment
Closure Type Snaps
Collar Type Lapel
Color Royal Blue
View More Specs
Products 11
Catalog Number Mfr. No. Size Price Quantity & Availability  
Catalog Number Mfr. No. Size Price Quantity & Availability  
18-111-316
View Documents
White Knight™
12L34940CXP63PXXSROY
2X-Small Each for $175.00
Add to cart
18-111-317
View Documents
White Knight™
12L34940CXP63PXSROY
X-Small Each for $175.00
Add to cart
18-111-318
View Documents
White Knight™
12L34940CXP63PSROY
Small Each for $175.00
Add to cart
18-111-319
View Documents
White Knight™
12L34940CXP63PMROY
Medium Each for $175.00
Add to cart
18-111-320
View Documents
White Knight™
12L34940CXP63PLROY
Large Each for $175.00
Add to cart
18-111-321
View Documents
White Knight™
12L34940CXP63PXLROY
X-Large Each for $185.00
Add to cart
18-111-322
View Documents
White Knight™
12L34940CXP63P2XLROY
2X-Large Each for $195.00
Add to cart
18-111-323
View Documents
White Knight™
12L34940CXP63P3XLROY
3X-Large Each for $210.00
Add to cart
18-111-324
View Documents
White Knight™
12L34940CXP63P4XLROY
4X-Large Each for $225.00
Add to cart
18-111-325
View Documents
White Knight™
12L34940CXP63P5XLROY
5X-Large Each for $248.00
Add to cart
18-111-326
View Documents
White Knight™
12L34940CXP63P6XLROY
6X-Large Each for $275.00
Add to cart
Description
Description
  • Best in class flammability protection
  • Reusable Nomex™ FR fabric
  • Meets NFPA 70E and HRC 1 performance standards
  • Quick-release snap front
  • Safety sleeve cuff with tunneled elastic
  • FR identification patches on sleeves and back
  • Excellent heat-resistance properties
Specifications
Specifications
HRC 1
Snaps
Royal Blue
No
Yes
3
NFPA 2112 Certified Garment
Lapel
Elastic
Unisex
Nomex™ CXP6 Style 106000
Non-sterile