Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Flame Tech Indicator 1, Reagents
Click to view available options
Quantity:
500 mL
Specifications
Specifications
Chemical Name or Material | Flame Tech Indicator 1 |
CAS | 64-17-5, 67-63-0, 67-56-1 |
Grade | Reagent |
Packaging | Glass Bottle |
Specific Gravity | 0.8 |
Color | Yellow |
Quantity | 500 mL |
Physical Form | Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction