Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human CELSR3 Alexa Fluor 488-conjugated Antibody, R&D Systems™
Mouse Monoclonal Antibody
Supplier: R&D Systems FAB7278G100UG
This item is not returnable.
View return policy
Description
Flamingo homolog 1/CELSR3 Monoclonal specifically detects Flamingo homolog 1/CELSR3 in Human samples. It is validated for Flow Cytometry.Specifications
Flamingo homolog 1/CELSR3 | |
Monoclonal | |
0.2 mg/mL | |
Flow Cytometry 0.25-1 ug/10^6 cells | |
Q9NYQ7 | |
CELSR3 | |
E. coli-derived recombinant human CELSR3 Val531-Ser711 Accession # Q9NYQ7 | |
RUO | |
1951 | |
Human | |
IgG2b |
Flow Cytometry | |
763103 | |
Alexa Fluor 488 | |
Supplied 0.2 mg/mL in a saline solution containing BSA and Sodium Azide. with 0.09% Sodium Azide | |
Cadherin family member 11, cadherin, EGF LAG seven-pass G-type receptor 3 (flamingo homolog, Drosophila), CDHF11, CDHF11RESDA1, EGFL1, EGFL1cadherin EGF LAG seven-pass G-type receptor 3, EGF-like protein 1, EGF-like-domain, multiple 1, epidermal growth factor-like 1, Epidermal growth factor-like protein 1, Flamingo homolog 1, FMI1anchor protein, HFMI1, KIAA0812, MEGF2, MEGF2cadherin EGF LAG seven-pass G-type receptor 3, flamingo (Drosophila) homolog, multiple EGF-like domains 2, Multiple EGF-like domains protein 2, Multiple epidermal growth factor-like domains protein 2, RESDA1 | |
Mouse | |
100 μg | |
Primary | |
Detects human CELSR3 in ELISA. | |
Store the unopened product at 2 - 8 degreesC. Do not use past expiration date. |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
For Research Use Only (RUO)
Spot an opportunity for improvement?
Provide Content Correction