Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific™ Flammable-Materials Storage Refrigerators Available on GSA/VA Contract for Federal Government customers only.

Ensure safe cold storage of volatile materials with flammable-materials refrigerators are that are engineered with no internal electrical components.

Manufacturer:  Thermo Scientific™ 20FREETSA

 View more versions of this product

Catalog No. 20FREETSA

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
View substitute products »


Description

Description

Ensure safe cold storage of volatile materials with Thermo Scientific™ Flammable-Materials Storage Refrigerators. These units are engineered with no internal electrical components that could trigger an explosion and meet OSHA and National Fire Protection Association safety requirements.

  • Temperature range: 2° to 7°C
  • Manual defrost
  • Hydraulic thermostat on exterior to eliminate interaction with stored materials
  • Aluminum evaporator coil
  • Magnetic vinyl gaskets for positive door seal
  • CFC-free insulation and coolant

This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.

Specifications

Specifications

Bottom Mounted
Polyurethane Foam
575 w
1°C to 12°C
20.0 cu. ft.
115 V 60 Hz
60 Hz
NEMA 5-15 P
23.8 x 27.8 x 57.8 in. (60.3 x 70.5 x 146.7 cm)
104.3 kg
R134a, 100% CFC Free
Door Lock
White
Indoor Cold Storage Of Flammable Solvents
3 in. On Sides, 3 in. At Back, 3 in. On Top
2 Adjustable Feet In Front, 2 Fixed Pads In Rear
Manual
Mechanical Dial Thermostat/Outside on Rear of Unit
Hinged, Opens left to Right, Non-Reversible
4 Fixed Shelves, 6 Fixed Door Shelves
Flammable Materials Storage (FMS) Refrigerator
cCSAus
566.34 L
115 V
5 A
32.13 x 34 x 73.13 in. (81.6 x 86.4 x 186.1 cm)
230 lb.
Temperature Display - None
Epoxy Coated Steel
13 Months Parts and Labor
Epoxy Coated Steel
10 Years
Interior - No
20FREETSA
Flammable Material Storage Refrigerator
SDS
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit