Learn More
Thermo Scientific™ Flammable-Materials Storage Refrigerators
Ensure safe cold storage of volatile materials with flammable-materials refrigerators that are engineered with no internal electrical components.
$11502.00
Specifications
Defrost Manual
View More Specs
Attention:This item requires special delivery information. You will be asked for this during the checkout process.
Products 4
Catalog Number Mfr. No. Description Electrical Requirements Plug Type Price Quantity & Availability  
Catalog Number Mfr. No. Description Electrical Requirements Plug Type Price Quantity & Availability  
50FREETSA
Accessories View Documents Promotion Details
Thermo Scientific™
50FREETSA
Flammable Materials Storage (FMS) Refrigerator 115 V 60 Hz NEMA 5-15 P
Each for $11,502.00
Add to cart
05FREETSA
Accessories View Documents Promotion Details
Thermo Scientific™
05FREETSA
Flammable Materials Storage (FMS) Refrigerator 115 V 60 Hz NEMA 5-15 P This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000. N/A
20FREETSA
Accessories View Documents Promotion Details
Thermo Scientific™
20FREETSA
Flammable Materials Storage (FMS) Refrigerator 115 V 60 Hz NEMA 5-15 P This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000. View substitute products »
20FRCETSA
Accessories View Documents Promotion Details
Thermo Scientific™
20FRCETSA
Corrosion Resistant, Flammable Materials Storage (FMS) Refrigerator 115 V 60 Hz NEMA 5-15 P This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000. N/A
Description
Description
Ensure safe cold storage of volatile materials with Thermo Scientific™ Flammable-Materials Storage Refrigerators. These units are engineered with no internal electrical components that could trigger an explosion and meet OSHA and National Fire Protection Association safety requirements.
  • Temperature range: 2° to 7°C
  • Manual defrost
  • Hydraulic thermostat on exterior to eliminate interaction with stored materials
  • Aluminum evaporator coil
  • Magnetic vinyl gaskets for positive door seal
  • CFC-free insulation and coolant
  • Warranty varies by country, please contact us for details
Warranty and Services
13 Months Parts and Labor
Specifications
Specifications
Manual