Learn More
Cole-Parmer Parker Hannifin Flaring tool for tubing 3/8" outer diameter  DFS Item
Manufacturer:  Cole-Parmer 3049005
Use these tools for a better fit. Heat the PFA tubing with a heat gun, and insert the tool to create the flare. Tools are available for any size of tubing from 1/4-inch to 1-inch outer diameter.
Catalog No. 50-632-003