Learn More
MilliporeSigma™ Supelco™ Flash Chromatography Assembly with Standard Column/Ball Joints, Joint Size: SJ 51/30
Flash chromatography is useful for rapid, preparative separations with moderate resolutions.
$770.00 - $848.50
Specifications
Column Type Flash Chromatography
For Use With (Application) Flash Chromatography
Joint Size SJ 51/30
View More Specs
Products 2
Catalog Number Mfr. No. Capacity (Metric) Diameter (Metric) Outer Length (Metric) Price Quantity & Availability  
Catalog Number Mfr. No. Capacity (Metric) Diameter (Metric) Outer Length (Metric) Price Quantity & Availability  
11-100-8443
View Documents
MilliporeSigma™ Supelco™
Z2023391EA
200 mL 25 mm 584 mm
Each for $770.00
Add to cart
11-100-8083
View Documents
MilliporeSigma™ Supelco™
Z2023981EA
400 mL 38 mm 560 mm
Each for $848.50
Add to cart
Description
Description
Consists of column, flow controller, and pinch clamp. Order solvent reservoir separately.
Specifications
Specifications
Flash Chromatography
Flash Chromatography
SJ 51/30