Learn More
BUCHI FlashPure EcoFlex Si LOK
Buchi FlashPure EcoFlex Si LOK is designed to optimized back pressure, price and performance. EcoFlex Silica is ideal for basic applications and polar compounds using organic solvents.
Manufacturer:  BUCHI 140000034
Catalog No. 05-410-016
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
Description
Description
  • 50 µm irregular particles
  • Luer Lok connection on both top and bottom of cartridge
  • Selection of cartridge sizes from 4 to 220 g
  • Non-RFID suitable on any system from BUCHI, ISCO, Biotage, Interchim or Yamazen.
Specifications
Specifications
Silica
25 g
12 Pc.