Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

BUCHI FlashPure EcoFlex Silica Lok
SDP

FlashPure EcoFlex cartridges are designed to optimized backpressure, price and performance. EcoFlex Silica is ideal for basic applications and polar compounds using organic solvents.

Manufacturer:  BUCHI 140000032

 View more versions of this product

Catalog No. 05-410-014

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.


Description

Description

  • 50 μm irregular particles
  • Most economical solution without sacrificing quality
  • Large selection of cartridge sizes from 4g-3000g
  • Non-RFID suitable on any system from BUCHI, ISCO, Biotage, Interchim or Yamazen.
Specifications

Specifications

Polypropylene
20 Pc.
4 g
Basic Applications, Polar Compounds
Silica
40 to 63 μm
Silica
Any Flash System
SDS
Documents
Product Certifications
Promotions

Promotions

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit