Learn More
BUCHI FlashPure EcoFlex Silica Solid Loader Insertion Rod
FlashPure EcoFlex Empty Solid Loaders are ideal for samples that are not readily soluble in the desired mobile phase when performing flash chromatogrpahy. The sample is adsorbed onto a solid phase (silica or Celite) and then added to the empty solid loader tube.
Manufacturer:  BUCHI 140000160
 View more versions of this product
Catalog No. 05-410-126
Description
Description
  • Screw cap makes sealing easy
  • Various sizes for differing sample amounts
  • Choose a size large enough for the sample
  • Insertion Rod allows frit placement and loading of samples smaller than the entire volume
Specifications
Specifications
Polypropylene
1 Pc.
Solid Loading of Insoluble Samples
Empty
12 to 40 g
Reveleris Solid Loading