Learn More
BUCHI FlashPure EcoFlex Silica Solid Loader Insertion Rod
FlashPure EcoFlex Empty Solid Loaders are ideal for samples that are not readily soluble in the desired mobile phase when performing flash chromatogrpahy. The sample is adsorbed onto a solid phase (silica or Celite) and then added to the empty solid loader tube.
$95.00 - $183.00
Specifications
For Use With (Application) Solid Loading of Insoluble Samples
For Use With (Equipment) Reveleris Solid Loading
Material Polypropylene
Type Empty
View More Specs
Products 4
Catalog Number Mfr. No. Size (Metric) Tube Rack Includes Price Quantity & Availability  
Catalog Number Mfr. No. Size (Metric) Tube Rack Includes Price Quantity & Availability  
05-410-126 View Documents BUCHI
140000160
12 to 40 g 1 Pc. Pack for $113.00
Add to cart
05-410-128 View Documents BUCHI
140000162
220 to 330 g 1 Pc. Pack for $183.00
Add to cart
05-410-125 View Documents BUCHI
140000159
4 g 1 Pc. Pack for $95.00
Add to cart
05-410-127 View Documents BUCHI
140000161
80 to 120 g 1 Pc. Pack for $146.00
Add to cart
Description
Description
  • Screw cap makes sealing easy
  • Various sizes for differing sample amounts
  • Choose a size large enough for the sample
  • Insertion Rod allows frit placement and loading of samples smaller than the entire volume
Specifications
Specifications
Solid Loading of Insoluble Samples
Reveleris Solid Loading
Polypropylene
Empty