Learn More
BUCHI FlashPure Empty Solid Loader
Buchi FlashPure Empty Solid Loader is ideal for samples that are not readily soluble in the desired mobile phase when performing flash chromatography. The sample is adsorbed onto a solid phase (silica or Celite) and then added to the empty solid loader tube.
Manufacturer:  BUCHI 140000158
Catalog No. 05-410-124
Add to cart
Description
Description
  • Screw cap makes sealing easy
  • Various sizes for differing sample amounts
  • Choose a size large enough for the sample
  • Insertion Rod allows frit placement and loading of samples smaller than the entire volume
Specifications
Specifications
120 g
5/Pk.