Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

BUCHI FlashPure ExoFlex Alumina Basic

FlashPure EcoFlex cartridges are designed to optimized backpressure, price and performance. EcoFlex Alumina Basic has the strongest retention properties for alcohols, hydrocarbons and steroids.

Manufacturer:  BUCHI 140000081

 View more versions of this product

Catalog No. 05-410-055


Add to cart

Description

Description

  • 60 µm irregular particles
  • Most economical solution without sacrificing quality
  • Large selection of cartridge sizes from 8g-3000g
  • Non-RFID suitable on any system from BUCHI, ISCO, Biotage, Interchim or Yamazen.
Specifications

Specifications

Alumina Basic
Any Flash System
Polypropylene
440 g
Polar Compounds
6 Pc.
50 to 75 μm
Basic Alumina
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit