Learn More
BUCHI FlashPure ExoFlex Alumina Basic
FlashPure EcoFlex cartridges are designed to optimized backpressure, price and performance. EcoFlex Alumina Basic has the strongest retention properties for alcohols, hydrocarbons and steroids.
Specifications
For Use With (Application) Polar Compounds
For Use With (Equipment) Any Flash System
Material Polypropylene
Type Basic Alumina
Column Type Alumina Basic
View More Specs
Products 5
Catalog Number Mfr. No. Size (Metric) Tube Rack Includes Price Quantity & Availability  
Catalog Number Mfr. No. Size (Metric) Tube Rack Includes Price Quantity & Availability  
05-410-053
View Documents
BUCHI
140000079
160 g 10 Pc. This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000. N/A
05-410-054
View Documents
BUCHI
140000080
240 g 10 Pc. This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000. N/A
05-410-051
View Documents
BUCHI
140000077
50 g 12 Pc. This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000. N/A
05-410-052
View Documents
BUCHI
140000078
80 g 12 Pc. This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000. N/A
05-410-049
View Documents
BUCHI
140000075
8 g 20 Pc. This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000. N/A
Description
Description
  • 60 μm irregular particles
  • Most economical solution without sacrificing quality
  • Large selection of cartridge sizes from 8g-3000g
  • Non-RFID suitable on any system from BUCHI, ISCO, Biotage, Interchim or Yamazen.
Specifications
Specifications
Polar Compounds
Polypropylene
Alumina Basic
Any Flash System
Basic Alumina