Learn More
BUCHI FlashPure EcoFlex Flash Chromatography Cartridges: Alumina Neutral

FlashPure EcoFlex cartridges are designed to optimized backpressure, price and performance.
$696.00 - $3853.00
Specifications
Material | Polypropylene |
---|---|
Type | Neutral Alumina |
Resolution | 50 to 75 μm |
Column Type | Alumina Neutral |
For Use With (Application) | Polar Compounds |
Catalog Number | Mfr. No. | Size (Metric) Tube Rack | Includes | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Size (Metric) Tube Rack | Includes | Price | Quantity & Availability | ||||
05-410-070
![]() |
BUCHI
140000096 |
24 g | 18 Pc. |
Pack of 18 for $696.00
|
|
||||
05-410-078
![]() |
BUCHI
140000104 |
3200 g | 2 Pc. |
Pack of 2 for $3,853.00
|
|
||||
05-410-069
![]() |
BUCHI
140000095 |
8 g | 20 Pc. | Item Discontinued This item has been discontinued by the supplier and is no longer available. Please reach out to your sales representative. | N/A | ||||
05-410-071
![]() |
BUCHI
140000097 |
50 g | 12 Pc. | Item Discontinued This item has been discontinued by the supplier and is no longer available. Please reach out to your sales representative. | N/A | ||||
05-410-072
![]() |
BUCHI
140000098 |
80 g | 12 Pc. | Item Discontinued This item has been discontinued by the supplier and is no longer available. Please reach out to your sales representative. | View substitute products » | ||||
Description
- Most economical solution without sacrificing quality for standard separations
- Large selection of cartridge sizes from 4g-5,000g
- Non-RFID suitable on any system from BUCHI, ISCO, Biotage, Interchim or Yamazen
- Silica is ideal for basic applications and polar compounds using organic solvents
- Amino functionalized silica provides better peak shapes with polar and basic compounds and is ideal for carbohydrates
- C18 functionalized silica is ideal for peptides and non-polar compounds in an aqueous mobile phase
- Diol functionalized silica provides an alternate selectivity for a variety of polarity types and is ideal for lipids
- Neutral Alumina is ideal for aldehydes, ketones, and esters
Specifications
Polypropylene | |
50 to 75 μm | |
Polar Compounds |
Neutral Alumina | |
Alumina Neutral | |
Any Flash System |
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.