Learn More
BUCHI FlashPure ID Amino Available on GSA/VA Contract for Federal Government customers only.
FlashPure ID cartridges are degined for Reveleris™ chromatography systems. Integrated RFID technology automatically transfers cartridge settings and data to the flash system. Amino functionalized silica provides better peak shapes with polar and basic compounds and is ideal for carbohydrates.
Manufacturer:  BUCHI 140000148
 View more versions of this product
Catalog No. 05-410-114
Add to cart
Description
Description
  • RFID for optimal convenience on BUCHI Reveleris™ systems
  • 40 µm irregular particle
  • Large selection of cartridge sizes from 4g-330g
This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.
Specifications
Specifications
Polypropylene
10 Pc.
220 g
Better Peak Shapes with Polar and Basic Compounds, Ideal for Carbohydrates
Amino
35 to 45 μm
Amino
Reveleris Flash System