Learn More
BUCHI FlashPure ID Amino Available on GSA/VA Contract for Federal Government customers only.
FlashPure ID cartridges are degined for Reveleris™ chromatography systems. Integrated RFID technology automatically transfers cartridge settings and data to the flash system. Amino functionalized silica provides better peak shapes with polar and basic compounds and is ideal for carbohydrates.
$182.00 - $13869.00
Specifications
For Use With (Application) Better Peak Shapes with Polar and Basic Compounds, Ideal for Carbohydrates
For Use With (Equipment) Reveleris Flash System
Material Polypropylene
Type Amino
Column Type Amino
View More Specs
Products 6
Catalog Number Mfr. No. Size (Metric) Tube Rack Includes Price Quantity & Availability  
Catalog Number Mfr. No. Size (Metric) Tube Rack Includes Price Quantity & Availability  
05-410-114 Available on GSA/VA Contract for Federal Government customers only. View Documents BUCHI
140000148
220 g 10 Pc. Pack of 10 for $8,674.00
Add to cart
05-410-111 Available on GSA/VA Contract for Federal Government customers only. View Documents BUCHI
140000145
220 g 1 Pc. Pack for $1,056.00
Add to cart
05-410-113 Available on GSA/VA Contract for Federal Government customers only. View Documents BUCHI
140000147
24 g 10 Pc. Pack of 10 for $1,626.00
Add to cart
05-410-000 Available on GSA/VA Contract for Federal Government customers only. View Documents BUCHI
140000017
24 g 1 Pc. Pack for $182.00
Add to cart
05-410-115 Available on GSA/VA Contract for Federal Government customers only. View Documents BUCHI
140000149
330 g 10 Pc. Pack of 10 for $13,869.00
Add to cart
05-410-112 Available on GSA/VA Contract for Federal Government customers only. View Documents BUCHI
140000146
330 g 1 Pc. Pack for $1,537.00
Add to cart
Description
Description
  • RFID for optimal convenience on BUCHI Reveleris™ systems
  • 40 µm irregular particle
  • Large selection of cartridge sizes from 4g-330g
This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.
Specifications
Specifications
Better Peak Shapes with Polar and Basic Compounds, Ideal for Carbohydrates
Polypropylene
Amino
Reveleris Flash System
Amino