Learn More
BUCHI FlashPure ID Amino Flash Cartridges PROMO
Better peak shapes with polar and basic compounds, ideal for carbohydrates
Manufacturer:  BUCHI 145159571PM
 View more versions of this product
Catalog No. 05-404-832PM
This item has been discontinued and is no longer available. Please call customer service for assistance: 1-800-766-7000.
Description
Description
High performance
  • High resolution of separation due to high-quality silica
  • Increased fraction purity due to a better separation
  • Faster evaporation of smaller fraction volumes
  • Reproducible performance from batch to batch
Lower purification costs
  • Increased sample throughput due to high sample loading capacity
  • Solvent saving: required purification yield is achieved faster
  • /ul>
Specifications
Specifications
12 g
10 Pc.
200 psig
Better Peak Shapes with Polar and Basic Compounds, Ideal for Carbohydrates
Silica - Amine
Reveleris