Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

BUCHI FlashPure ID C18 Flash Cartridges PROMO

Effective for non-polar compounds

Specifications

For Use With (Application) Effective for Non-Polar Compounds
Material Silica - C18
Max. Pressure 200 psig
Includes 1 Pc.
Type Reveleris
View More Specs

Products 4
Catalog Number Mfr. No. Bed Weight (Metric) Price Quantity & Availability  
Catalog Number Mfr. No. Bed Weight (Metric) Price Quantity & Availability  
05-404-807PM View Documents BUCHI
145152104PM
40 g This item has been discontinued and is no longer available. Please call customer service for assistance: 1-800-766-7000.
View substitute products »
05-404-808PM View Documents BUCHI
145152105PM
80 g This item has been discontinued and is no longer available. Please call customer service for assistance: 1-800-766-7000.
View substitute products »
05-404-806PM View Documents BUCHI
145152103PM
12 g This item has been discontinued and is no longer available. Please call customer service for assistance: 1-800-766-7000.
View substitute products »
05-404-810PM View Documents BUCHI
145152991PM
120 g This item has been discontinued and is no longer available. Please call customer service for assistance: 1-800-766-7000.
View substitute products »
Description

Description

High performance

  • High resolution of separation due to high-quality silica
  • Increased fraction purity due to a better separation
  • Faster evaporation of smaller fraction volumes
  • Reproducible performance from batch to batch

Lower purification costs

  • Increased sample throughput due to high sample loading capacity
  • Solvent saving: required purification yield is achieved faster
  • /ul>
Specifications

Specifications

Effective for Non-Polar Compounds
200 psig
Reveleris
Silica - C18
1 Pc.
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit