Learn More
BUCHI FlashPure ID C18-WP Chromatography Cartridges
FlashPure ID cartridges are designed for BUCHI Pure chromatography systems. Integrated RFID technology automatically transfers cartridge settings and data to the flash system.
Manufacturer:  BUCHI 140000019
 View more versions of this product
Catalog No. 05-410-002
Add to cart
Description
Description
  • High resolution of separation due to high-quality silica
  • Increased fraction purity due to a better separation
  • Faster evaporation of smaller fraction volumes
  • Reproducible performance from batch to batch
  • Increased sample throughput due to high sample loading capacity
  • Solvent saving: required purification yield is achieved faster
Specifications
Specifications
Polypropylene
1 Pc.
24 g
Wide Pore Media for Large Molecules, Ideal for Peptides, Protiens and Lipids
C18 Wide Pore
16 to 24 μm
C18-WP
Reveleris Flash System