Learn More
BUCHI FlashPure ID C18-WP
FlashPure ID cartridges are degined for Reveleris™ chromatography systems. Integrated RFID technology automatically transfers cartridge settings and data to the flash system. C18-WP (Wide Pore) is ideal for large molecules, peptides, proteins and lipids.
Manufacturer:  BUCHI 140000152
 View more versions of this product
Catalog No. 05-410-118
Add to cart
Description
Description
  • RFID for optimal convenience on BUCHI Reveleris™ systems
  • 20 µm irregular particle for difficult separations
  • Wide pore silica gel for large molecule separations
  • Large selection of cartridge sizes from 4g-330g
Specifications
Specifications
Polypropylene
1 Pc.
80 g
Wide Pore Media for Large Molecules, Ideal for Peptides, Protiens and Lipids
C18 Wide Pore
16 to 24 μm
C18-WP
Reveleris Flash System