Learn More
BUCHI FlashPure ID C18-WP Chromatography Cartridges Available on GSA/VA Contract for Federal Government customers only.
FlashPure ID cartridges are designed for BUCHI Pure chromatography systems. Integrated RFID technology automatically transfers cartridge settings and data to the flash system.
Manufacturer:  BUCHI 140000154
 View more versions of this product
Catalog No. 05-410-120
Add to cart
Description
Description
  • High resolution of separation due to high-quality silica
  • Increased fraction purity due to a better separation
  • Faster evaporation of smaller fraction volumes
  • Reproducible performance from batch to batch
  • Increased sample throughput due to high sample loading capacity
  • Solvent saving: required purification yield is achieved faster
This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.
Specifications
Specifications
Polypropylene
1 Pc.
220 g
Wide Pore Media for Large Molecules, Ideal for Peptides, Protiens and Lipids
C18 Wide Pore
16 to 24 μm
C18-WP
Reveleris Flash System