Learn More
BUCHI FlashPure ID C18-WP Available on GSA/VA Contract for Federal Government customers only.
FlashPure ID cartridges are degined for Reveleris™ chromatography systems. Integrated RFID technology automatically transfers cartridge settings and data to the flash system. C18-WP (Wide Pore) is ideal for large molecules, peptides, proteins and lipids.
Manufacturer:  BUCHI 140000155
 View more versions of this product
Catalog No. 05-410-121
Add to cart
Description
Description
  • RFID for optimal convenience on BUCHI Reveleris™ systems
  • 20 µm irregular particle for difficult separations
  • Wide pore silica gel for large molecule separations
  • Large selection of cartridge sizes from 4g-330g
This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.
Specifications
Specifications
Polypropylene
1 Pc.
330 g
Wide Pore Media for Large Molecules, Ideal for Peptides, Protiens and Lipids
C18 Wide Pore
16 to 24 μm
C18-WP
Reveleris Flash System