Learn More
BUCHI FlashPure ID C18-WP Available on GSA/VA Contract for Federal Government customers only.
FlashPure ID cartridges are degined for Reveleris™ chromatography systems. Integrated RFID technology automatically transfers cartridge settings and data to the flash system. C18-WP (Wide Pore) is ideal for large molecules, peptides, proteins and lipids.
$214.00 - $1739.00
Specifications
For Use With (Application) Wide Pore Media for Large Molecules, Ideal for Peptides, Protiens and Lipids
For Use With (Equipment) Reveleris Flash System
Material Polypropylene
Type C18 Wide Pore
Column Type C18-WP
View More Specs
Products 5
Catalog Number Mfr. No. Size (Metric) Tube Rack Includes Price Quantity & Availability  
Catalog Number Mfr. No. Size (Metric) Tube Rack Includes Price Quantity & Availability  
05-410-119 Available on GSA/VA Contract for Federal Government customers only.
View Documents
BUCHI
140000153
120 g 1 Pc. Pack for $652.00
Add to cart
05-410-120 Available on GSA/VA Contract for Federal Government customers only.
View Documents
BUCHI
140000154
220 g 1 Pc. Pack for $1,221.00
Add to cart
05-410-002 Available on GSA/VA Contract for Federal Government customers only.
View Documents
BUCHI
140000019
24 g 1 Pc. Pack for $214.00
Add to cart
05-410-121 Available on GSA/VA Contract for Federal Government customers only.
View Documents
BUCHI
140000155
330 g 1 Pc. Pack for $1,739.00
Add to cart
05-410-118 Available on GSA/VA Contract for Federal Government customers only.
View Documents
BUCHI
140000152
80 g 1 Pc. Pack for $414.00
Add to cart
Description
Description
  • RFID for optimal convenience on BUCHI Reveleris™ systems
  • 20 µm irregular particle for difficult separations
  • Wide pore silica gel for large molecule separations
  • Large selection of cartridge sizes from 4g-330g
This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.
Specifications
Specifications
Wide Pore Media for Large Molecules, Ideal for Peptides, Protiens and Lipids
Polypropylene
C18-WP
Reveleris Flash System
C18 Wide Pore