Learn More
BUCHI FlashPure ID C18-WP Flash Cartridges PROMO
Ideal for peptides, proteins and lipids
Manufacturer:  BUCHI 145169483PM
 View more versions of this product
Catalog No. 05-404-840PM
This item has been discontinued and is no longer available. Please call customer service for assistance: 1-800-766-7000.
View substitute products »
Description
Description
High performance
  • High resolution of separation due to high-quality silica
  • Increased fraction purity due to a better separation
  • Faster evaporation of smaller fraction volumes
  • Reproducible performance from batch to batch
Lower purification costs
  • Increased sample throughput due to high sample loading capacity
  • Solvent saving: required purification yield is achieved faster
  • /ul>
Specifications
Specifications
12 g
1 Pc.
200 psig
Ideal for Peptides, Proteins and Lipids and Other Large Molecule Purifications
Silica - C-18 WP
Reveleris